siRNA Details
| Gene Symbol: Rab22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024436.3 | NP_077756.2 | 4 | 507 | ||
| GenBank mRNA | AB232616.1 | - | 4 | 246 | ||
| AJ311124.1 | - | 4 | 303 | |||
| AK014407.1 | - | 493 | ||||
| AK031667.1 | - | 4 | 487 | |||
| AK146594.1 | - | 4 | 507 | |||
| AK147619.1 | - | 511 | ||||
| AK149200.1 | - | 499 | ||||
| AK150617.1 | - | 4 | 513 | |||
| AK219950.1 | - | 4 | 252 | |||
| BC006596.1 | - | 4 | 494 | 3965264 | ||

Rab22a
Genomic Map