siRNA Details
| Gene Symbol: Rab22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024436.3 | NP_077756.2 | 7 | 1175 | ||
| GenBank mRNA | AJ311124.1 | - | 7 | 971 | ||
| AK014407.1 | - | 1161 | ||||
| AK031667.1 | - | 7 | 1155 | |||
| AK146594.1 | - | 7 | 1175 | |||
| AK147619.1 | - | 1179 | ||||
| AK149200.1 | - | 1168 | ||||
| AK150617.1 | - | 7 | 1181 | |||
| BC006596.1 | - | 7 | 1162 | 3965264 | ||

Rab22a
Genomic Map