siRNA Details
| Gene Symbol: Krtcap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025327.2 | NP_079603.1 | 4 | 352 | ||
| GenBank mRNA | AK002479.1 | - | 4 | 353 | ||
| AK003310.1 | - | 4 | 320 | |||
| BC043030.1 | - | 4 | 824 | 5693076 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtcap2| Gene Symbol: Krtcap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025327.2 | NP_079603.1 | 4 | 352 | ||
| GenBank mRNA | AK002479.1 | - | 4 | 353 | ||
| AK003310.1 | - | 4 | 320 | |||
| BC043030.1 | - | 4 | 824 | 5693076 | ||