siRNA Details
| Gene Symbol: Chchd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025336.1 | NP_079612.1 | 8 | 1127 | ||
| GenBank mRNA | AK002889.1 | - | 8 | 1199 | ||
| AK003519.1 | - | 8 | 1127 | |||
| BC032987.1 | - | 8 | 1142 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chchd3| Gene Symbol: Chchd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025336.1 | NP_079612.1 | 8 | 1127 | ||
| GenBank mRNA | AK002889.1 | - | 8 | 1199 | ||
| AK003519.1 | - | 8 | 1127 | |||
| BC032987.1 | - | 8 | 1142 | |||