siRNA Details
| Gene Symbol: Mrps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025369.3 | NP_079645.1 | 2, 3 | 190 | ||
| GenBank mRNA | AK003111.1 | - | 2, 3 | 120 | ||
| AK003765.1 | - | 2, 3 | 167 | |||
| AK007512.1 | - | 2, 3 | 161 | |||
| AK009050.1 | - | 2, 3 | 169 | |||
| BC028499.1 | - | 2, 3 | 146 | 1382215 | ||
| BC096576.1 | - | 2, 3 | 155 | 1401014 | ||

Mrps36
Genomic Map