siRNA Details
| Gene Symbol: Fbxo36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025386.3 | NP_079662.1 | 4 | 780 | ||
| GenBank mRNA | AK002325.1 | - | 4 | 760 | ||
| AK003856.1 | - | 4 | 780 | |||
| AK008324.1 | - | 4 | 777 | |||
| AK010335.1 | - | 4 | 1593 | |||
| BC049581.1 | - | 4 | 757 | 6772363 | ||

Fbxo36
Genomic Map