siRNA Details
| Gene Symbol: Tpd52l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291197.1 | NP_001278126.1 | 8 | 1767 | ||
| NM_001291200.1 | NP_001278129.1 | 7 | 1722 | |||
| NM_001291201.1 | NP_001278130.1 | 7 | 1707 | |||
| NM_001291202.1 | NP_001278131.1 | 7 | 1704 | |||
| NM_001291203.1 | NP_001278132.1 | 6 | 1662 | |||
| NM_001291204.1 | NP_001278133.1 | 5 | 1513 | |||
| NM_025482.3 | NP_079758.2 | 8 | 1764 | |||
| GenBank mRNA | AK051268.1 | - | 7 | 1648 | ||
| AK147885.1 | - | 7 | 1682 | |||
| AK161092.1 | - | 6 | 1618 | |||
| BC006886.1 | - | 8 | 1653 | 2647702 | ||

Tpd52l2
Genomic Map