siRNA Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160208.1 | NP_001153680.1 | 6 | 1071 | ||
| NM_001160209.1 | NP_001153681.1 | 5 | 1158 | |||
| NM_025505.4 | NP_079781.2 | 7 | 1092 | |||
| GenBank mRNA | AK005489.1 | - | 5 | 892 | ||
| AK006544.1 | - | 6 | 1075 | |||
| AK029787.1 | - | 7 | 1093 | |||
| AK167493.1 | - | 5 | 1165 | |||
| AK168933.1 | - | 5 | 919 | |||
| AK171167.1 | - | 5 | 950 | |||
| BC027025.1 | - | 5 | 933 | 5374749 | ||

Blzf1
Genomic Map