siRNA Details
| Gene Symbol: Rtca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025517.3 | NP_079793.2 | 6 | 780 | ||
| GenBank mRNA | AK002543.1 | - | 6 | 690 | ||
| AK009245.1 | - | 6 | 716 | |||
| AK151108.1 | - | 6 | 668 | |||
| AK151400.1 | - | 6 | 668 | |||
| AK152129.1 | - | 6 | 781 | |||
| AK159961.1 | - | 6 | 670 | |||
| AK167830.1 | - | 6 | 657 | |||
| BC016519.1 | - | 6 | 706 | 3495638 | ||

Rtca
Genomic Map