siRNA Details
| Gene Symbol: Rtca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025517.3 | NP_079793.2 | 11 | 1241 | ||
| GenBank mRNA | AK002543.1 | - | 11 | 1151 | ||
| AK009245.1 | - | 11 | 1177 | |||
| AK152129.1 | - | 11 | 1242 | |||
| AK159961.1 | - | 11 | 1131 | |||
| AK167830.1 | - | 11 | 1118 | |||
| BC042473.1 | - | 2 | 3624 | |||
| BC051062.1 | - | 2 | 3624 | |||
| BC090403.1 | - | 11 | 1212 | 5293337 | ||

Rtca
Genomic Map