siRNA Details
| Gene Symbol: Sar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025535.2 | NP_079811.1 | 7 | 1059 | ||
| GenBank mRNA | AK002327.1 | - | 7 | 1036 | ||
| AK010187.1 | - | 7 | 1042 | |||
| AK013559.1 | - | 7 | 1021 | |||
| AK155429.1 | - | 7 | 1029 | |||
| BB749343.1 | - | 1 | 241 | |||
| BC082550.1 | - | 7 | 1028 | 30357800 | ||
| BC085178.1 | - | 7 | 1012 | 30701201 | ||

Sar1b
Genomic Map