siRNA Details
Gene Symbol: Commd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025536.2 | NP_079812.1 | 2 | 253 | ||
GenBank mRNA | AK010045.1 | - | 2 | 254 | ||
AK013148.1 | - | 1 | 638 | |||
BC025891.1 | - | 2 | 218 | 5035059 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
