siRNA Details
| Gene Symbol: Farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025648.3 | NP_079924.2 | 4, 5 | 546 | ||
| GenBank mRNA | AK002577.1 | - | 4, 5 | 523 | ||
| AK016506.1 | - | 4, 5 | 542 | |||
| AK031697.1 | - | 4, 5 | 524 | |||
| AK044637.1 | - | 4, 5 | 524 | |||
| AK084031.1 | - | 4, 5 | 508 | |||
| AK154629.1 | - | 4, 5 | 544 | |||
| AK171872.1 | - | 4, 5 | 427 | |||
| BC006862.1 | - | 4, 5 | 517 | 3595108 | ||
| BC013533.1 | - | 4, 5 | 515 | 4238911 | ||
| CT010350.1 | - | 4, 5 | 497 | |||

Farsa
Genomic Map