siRNA Details
| Gene Symbol: Mcm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291054.1 | NP_001277983.1 | 19 | 2973 | ||
| NM_025676.4 | NP_079952.2 | 18 | 2889 | |||
| GenBank mRNA | AK133858.1 | - | 19 | 2930 | ||
| BC052070.1 | - | 18 | 2695 | 5716125 | ||

Mcm8
Genomic Map