siRNA Details
| Gene Symbol: Bcl2l14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025778.3 | NP_080054.1 | 7 | 1556 | ||
| GenBank mRNA | AK008682.1 | - | 6 | 1481 | ||
| AK016670.1 | - | 7 | 1557 | |||
| AK018579.1 | - | 6 | 1523 | |||
| BC025541.1 | - | 6 | 1473 | 5253808 | ||

Bcl2l14
Genomic Map