siRNA Details
| Gene Symbol: Tax1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025816.3 | NP_080092.2 | 4 | 459 | ||
| GenBank mRNA | AK004574.1 | - | 4 | 441 | ||
| AK146076.1 | - | 4 | 439 | |||
| AK171136.1 | - | 4 | 426 | |||
| BC014798.1 | - | 4 | 380 | 3962810 | ||

Tax1bp1
Genomic Map