siRNA Details
| Gene Symbol: Cdca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025866.3 | NP_080142.1 | 5, 6 | 763 | ||
| GenBank mRNA | AK011289.1 | - | 5, 6 | 764 | ||
| AK028671.1 | - | 5, 6 | 774 | |||
| AK075882.1 | - | 5, 6 | 728 | |||
| AK153943.1 | - | 5, 6 | 742 | |||
| BC066169.1 | - | 5, 6 | 771 | 30474357 | ||

Cdca7
Genomic Map