siRNA Details
| Gene Symbol: Prl3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025896.2 | NP_080172.1 | 4, 5 | 644 | ||
| GenBank mRNA | AF525154.1 | - | 4, 5 | 629 | ||
| AK005474.1 | - | 4, 5 | 645 | |||
| AK005567.1 | - | 4, 5 | 642 | |||
| BC100334.1 | - | 4, 5 | 581 | 30791042 | ||

Prl3a1
Genomic Map