siRNA Details
| Gene Symbol: Zbtb8os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025970.3 | NP_080246.1 | 7 | 457 | ||
| GenBank mRNA | AK007377.1 | - | 7 | 447 | ||
| AK008007.1 | - | 7 | 458 | |||
| AK132279.1 | - | 468 | ||||
| AY071852.1 | - | 7 | 446 | |||
| BC026442.1 | - | 452 | ||||
| BC037657.1 | - | 427 | ||||
| BC046825.1 | - | 464 | ||||
| BC094629.1 | - | 7 | 488 | 3376726 | ||
| BC125606.1 | - | 7 | 458 | 40130121 | ||
| BC125608.1 | - | 7 | 458 | 40130123 | ||

Zbtb8os
Genomic Map