siRNA Details
| Gene Symbol: Tomm34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291155.1 | NP_001278084.1 | 7 | 1709 | ||
| NM_025996.5 | NP_080272.1 | 7 | 2088 | |||
| GenBank mRNA | AK011786.1 | - | 7 | 2088 | ||
| AK017699.1 | - | 7 | 1654 | |||
| AK075873.1 | - | 7 | 1707 | |||
| AK144450.1 | - | 4 | 1877 | |||
| AK168329.1 | - | 7 | 1644 | |||
| BC018278.1 | - | 7 | 1639 | 4187356 | ||

Tomm34
Genomic Map