siRNA Details
| Gene Symbol: LRRC32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128922.1 | NP_001122394.1 | 3 | 763 | ||
| NM_005512.2 | NP_005503.1 | 3 | 868 | |||
| XM_005273902.3 | XP_005273959.1 | 3 | 957 | |||
| XM_005273903.4 | XP_005273960.2 | 3 | 850 | |||
| XM_011544935.2 | XP_011543237.1 | 3 | 1159 | |||
| XM_017017527.1 | XP_016873016.1 | 3 | 1363 | |||
| GenBank mRNA | AK314971.1 | - | 3 | 842 | ||
| BC052210.1 | - | 3 | 760 | 4825609 | ||
| BC070079.1 | - | 3 | 763 | 30344529 | ||
| BI010671.1 | - | 1 | 492 | |||
| Z24680.1 | - | 3 | 719 | |||

LRRC32
Genomic Map