siRNA Details
| Gene Symbol: Prl7c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026206.2 | NP_080482.1 | 6 | 912 | ||
| GenBank mRNA | AK005486.1 | - | 6 | 913 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl7c1| Gene Symbol: Prl7c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026206.2 | NP_080482.1 | 6 | 912 | ||
| GenBank mRNA | AK005486.1 | - | 6 | 913 | ||