siRNA Details
| Gene Symbol: Ctdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026295.2 | NP_080571.2 | 8 | 1950 | ||
| GenBank mRNA | AK156617.1 | - | 8 | 1979 | ||
| BC053435.1 | - | 8 | 1950 | 6394578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctdp1| Gene Symbol: Ctdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026295.2 | NP_080571.2 | 8 | 1950 | ||
| GenBank mRNA | AK156617.1 | - | 8 | 1979 | ||
| BC053435.1 | - | 8 | 1950 | 6394578 | ||