siRNA Details
| Gene Symbol: Mphosph10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026483.2 | NP_080759.2 | 7 | 1392 | ||
| GenBank mRNA | AK008785.1 | - | 4 | 322 | ||
| AK013337.1 | - | 7 | 1380 | |||
| BC018177.1 | - | 7 | 1386 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mphosph10| Gene Symbol: Mphosph10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026483.2 | NP_080759.2 | 7 | 1392 | ||
| GenBank mRNA | AK008785.1 | - | 4 | 322 | ||
| AK013337.1 | - | 7 | 1380 | |||
| BC018177.1 | - | 7 | 1386 | |||