siRNA Details
| Gene Symbol: Nacc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037098.1 | NP_001032175.1 | 6 | 2515 | ||
| NM_026495.3 | NP_080771.3 | 6 | 1992 | |||
| GenBank mRNA | AK002651.1 | - | 6 | 1966 | ||
| AK002942.1 | - | 1 | 151 | |||
| AK147293.1 | - | 6 | 1992 | |||
| AK147310.1 | - | 6 | 2096 | |||
| AK187816.1 | - | 1 | 170 | |||
| BC022103.1 | - | 6 | 1974 | 5067063 | ||
| BC070453.1 | - | 6 | 2295 | |||

Nacc2
Genomic Map