siRNA Details
| Gene Symbol: Txndc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026559.3 | NP_080835.1 | 1 | 173 | ||
| GenBank mRNA | AJ344103.1 | - | 1 | 162 | ||
| AK004219.1 | - | 1 | 181 | |||
| AK007595.1 | - | 1 | 177 | |||
| AK013103.1 | - | 1 | 116 | |||
| AK076425.1 | - | 1 | 173 | |||
| AK169882.1 | - | 1 | 116 | |||
| BC030344.1 | - | 1 | 171 | 3673521 | ||

Txndc17
Genomic Map