siRNA Details
| Gene Symbol: Lrrc16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026825.3 | NP_081101.3 | 28 | 2633 | ||
| GenBank mRNA | AK043591.1 | - | 13 | 1213 | ||
| AK049059.1 | - | 10 | 1009 | |||
| AY437876.1 | - | 28 | 2638 | |||
| BC012229.1 | - | 7 | 662 | |||
| BC150819.1 | - | 27 | 2547 | |||

Lrrc16a
Genomic Map