siRNA Details
| Gene Symbol: Lrrc16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026825.3 | NP_081101.3 | 33 | 3309 | ||
| GenBank mRNA | AK043591.1 | - | 19 | 1907 | ||
| AY437876.1 | - | 33 | 3314 | |||
| BC012229.1 | - | 12 | 1338 | |||
| BC150819.1 | - | 32 | 3223 | |||

Lrrc16a
Genomic Map