siRNA Details
| Gene Symbol: Zc3h15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026934.3 | NP_081210.2 | 8 | 962 | ||
| GenBank mRNA | AK007452.1 | - | 3 | 193 | ||
| AK139511.1 | - | 8 | 1058 | |||
| AK152374.1 | - | 8 | 1053 | |||
| AK159596.1 | - | 8 | 1034 | |||
| AK167696.1 | - | 8 | 1055 | |||
| BC028787.1 | - | 3 | 401 | 1246186 | ||
| BC031845.1 | - | 8 | 962 | 3983096 | ||
| BC058229.1 | - | 8 | 889 | 6313954 | ||

Zc3h15
Genomic Map