siRNA Details
| Gene Symbol: Mospd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290514.1 | NP_001277443.1 | 4 | 648 | ||
| NM_027409.5 | NP_081685.1 | 4 | 648 | |||
| GenBank mRNA | AK007533.1 | - | 4 | 581 | ||
| AK088275.1 | - | 4 | 585 | |||
| AK139795.1 | - | 4 | 592 | |||
| BC018329.1 | - | 4 | 549 | 3962906 | ||
| BC085100.1 | - | 4 | 556 | 6391017 | ||
| BC100442.1 | - | 4 | 536 | 30501889 | ||

Mospd1
Genomic Map