siRNA Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027547.2 | NP_081823.2 | 15 | 1960 | ||
| GenBank mRNA | AK014501.1 | - | 15 | 1884 | ||
| AK018310.1 | - | 11 | 1346 | |||
| AK021384.1 | - | 6 | 769 | |||
| AK031424.1 | - | 1 | 1065 | |||
| BX511446.1 | - | 4 | 463 | |||

Prdm5
Genomic Map