siRNA Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027547.2 | NP_081823.2 | 15 | 1969 | ||
| GenBank mRNA | AK014501.1 | - | 15 | 1893 | ||
| AK018310.1 | - | 11 | 1355 | |||
| AK021384.1 | - | 6 | 778 | |||
| AK031424.1 | - | 1 | 1074 | |||
| BX511446.1 | - | 4 | 472 | |||

Prdm5
Genomic Map