siRNA Details
| Gene Symbol: Clec2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168223.1 | NP_001161695.1 | 6 | 1290 | ||
| NM_001168224.1 | NP_001161696.1 | 6 | 1402 | |||
| NM_027562.4 | NP_081838.1 | 7 | 1464 | |||
| GenBank mRNA | AK014570.1 | - | 7 | 1466 | ||
| AK137753.1 | - | 6 | 1400 | |||

Clec2g
Genomic Map