siRNA Details
| Gene Symbol: Adam30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027665.1 | NP_081941.1 | 1 | 334 | ||
| GenBank mRNA | AK016888.1 | - | 2 | 117 | ||
| AK077058.1 | - | 487 | ||||
| AY190760.1 | - | 1 | 334 | |||
| BC113152.1 | - | 1 | 326 | 40059344 | ||
| BC113762.1 | - | 1 | 326 | 40059342 | ||

Adam30
Genomic Map