siRNA Details
| Gene Symbol: Slc16a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027921.1 | NP_082197.1 | 4 | 1318 | ||
| GenBank mRNA | AK003423.1 | - | 1 | 562 | ||
| AK049846.1 | - | 4 | 1370 | |||
| BC023456.1 | - | 4 | 1318 | 5041164 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a14| Gene Symbol: Slc16a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027921.1 | NP_082197.1 | 4 | 1318 | ||
| GenBank mRNA | AK003423.1 | - | 1 | 562 | ||
| AK049846.1 | - | 4 | 1370 | |||
| BC023456.1 | - | 4 | 1318 | 5041164 | ||