siRNA Details
| Gene Symbol: Zswim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027964.2 | NP_082240.1 | 4 | 408 | ||
| GenBank mRNA | AK006367.1 | - | 4 | 409 | ||
| AK017097.1 | - | 4 | 409 | |||
| BC120837.1 | - | 4 | 388 | 40129760 | ||
| BC144860.1 | - | 4 | 388 | |||

Zswim2
Genomic Map