siRNA Details
| Gene Symbol: Epn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027984.3 | NP_082260.1 | 2 | 863 | ||
| GenBank mRNA | AK009469.1 | - | 1 | 243 | ||
| AK158031.1 | - | 2 | 822 | |||
| BC016454.1 | - | 2 | 812 | 4217221 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epn3| Gene Symbol: Epn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027984.3 | NP_082260.1 | 2 | 863 | ||
| GenBank mRNA | AK009469.1 | - | 1 | 243 | ||
| AK158031.1 | - | 2 | 822 | |||
| BC016454.1 | - | 2 | 812 | 4217221 | ||