siRNA Details
| Gene Symbol: Fbxo22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028049.2 | NP_082325.2 | 5, 6 | 737 | ||
| GenBank mRNA | AK002745.1 | - | 5, 6 | 617 | ||
| AK005473.1 | - | 4, 5 | 565 | |||
| AK182889.1 | - | 1, 2 | 172 | |||
| AK188724.1 | - | 1, 2 | 172 | |||
| AK207864.1 | - | 1, 2 | 172 | |||
| AK211102.1 | - | 1, 2 | 39 | |||
| AK213133.1 | - | 1, 2 | 172 | |||
| AK219248.1 | - | 1, 2 | 172 | |||
| BC018273.1 | - | 5, 6 | 677 | 4458296 | ||

Fbxo22
Genomic Map