siRNA Details
| Gene Symbol: C1qtnf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204152.1 | NP_001191081.1 | 2 | 292 | ||
| NM_001204153.1 | NP_001191082.1 | 2 | 292 | |||
| NM_028331.5 | NP_082607.3 | 2 | 292 | |||
| GenBank mRNA | AK012868.1 | - | 2 | 234 | ||
| AK051058.1 | - | 2 | 206 | |||
| AK154407.1 | - | 2 | 269 | |||
| AK198051.1 | - | 1 | 17 | |||
| BC025489.1 | - | 2 | 235 | |||
| BC071187.1 | - | 2 | 259 | 6395221 | ||
| DQ002399.1 | - | 2 | 258 | |||

C1qtnf6
Genomic Map