siRNA Details
| Gene Symbol: C1qtnf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204152.1 | NP_001191081.1 | 4 | 1561 | ||
| NM_001204153.1 | NP_001191082.1 | 3 | 1516 | |||
| NM_028331.5 | NP_082607.3 | 3 | 1727 | |||
| GenBank mRNA | AK012868.1 | - | 4 | 1503 | ||
| AK051058.1 | - | 3 | 1641 | |||
| AK154407.1 | - | 3 | 1704 | |||
| BC025489.1 | - | 3 | 1459 | |||
| BC071187.1 | - | 3 | 1694 | 6395221 | ||

C1qtnf6
Genomic Map