siRNA Details
| Gene Symbol: 4933414I15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016566.1 | - | 1 | 2426 | ||
| AK016600.1 | - | 2 | 1587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933414I15Rik| Gene Symbol: 4933414I15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016566.1 | - | 1 | 2426 | ||
| AK016600.1 | - | 2 | 1587 | |||