siRNA Details
| Gene Symbol: Ccdc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029212.3 | NP_083488.1 | 5, 6 | 828 | ||
| GenBank mRNA | AB048860.1 | - | 5, 6 | 902 | ||
| BC116291.1 | - | 5, 6 | 828 | 40094912 | ||
| BC116292.1 | - | 5, 6 | 828 | 40094918 | ||

Ccdc33
Genomic Map