siRNA Details
| Gene Symbol: Ccdc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB048860.1 | - | 2629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc33| Gene Symbol: Ccdc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB048860.1 | - | 2629 | |||