siRNA Details
| Gene Symbol: Gpsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029522.2 | NP_083798.2 | 3, 4 | 792 | ||
| GenBank mRNA | AK040996.1 | - | 3, 4 | 681 | ||
| AK143552.1 | - | 3, 4 | 793 | |||
| AY081187.1 | - | 3, 4 | 636 | |||
| BC021308.1 | - | 2, 3 | 347 | 5007832 | ||

Gpsm2
Genomic Map