siRNA Details
| Gene Symbol: Gpsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029522.2 | NP_083798.2 | 5, 6 | 1073 | ||
| GenBank mRNA | AK040996.1 | - | 5, 6 | 962 | ||
| AK143552.1 | - | 5, 6 | 1074 | |||
| AY081187.1 | - | 5, 6 | 917 | |||
| BC021308.1 | - | 4, 5 | 628 | 5007832 | ||

Gpsm2
Genomic Map