siRNA Details
| Gene Symbol: Depdc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172092.1 | NP_001165563.1 | 8 | 1576 | ||
| NM_029523.3 | NP_083799.2 | 8 | 1576 | |||
| GenBank mRNA | AK018060.1 | - | 1 | 99 | ||
| AK146458.1 | - | 8 | 1545 | |||
| BC023934.1 | - | 8 | 1569 | 5342167 | ||

Depdc1a
Genomic Map