siRNA Details
| Gene Symbol: Lbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029999.4 | NP_084275.3 | 3 | 1302 | ||
| GenBank mRNA | AF317517.1 | - | 3 | 1283 | ||
| BC029988.1 | - | 1 | 433 | |||
| BC052470.1 | - | 3 | 1282 | 6813866 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lbh| Gene Symbol: Lbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029999.4 | NP_084275.3 | 3 | 1302 | ||
| GenBank mRNA | AF317517.1 | - | 3 | 1283 | ||
| BC029988.1 | - | 1 | 433 | |||
| BC052470.1 | - | 3 | 1282 | 6813866 | ||