siRNA Details
| Gene Symbol: Rbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030081.2 | NP_084357.2 | 5, 6 | 711 | ||
| GenBank mRNA | AK019909.2 | - | 5, 6 | 750 | ||
| BC017622.1 | - | 5, 6 | 758 | |||
| BC031135.1 | - | 4, 5 | 542 | 4971294 | ||
| BC049106.1 | - | 5, 6 | 711 | 6412839 | ||

Rbsn
Genomic Map