siRNA Details
| Gene Symbol: Clip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271483.1 | NP_001258412.1 | 4 | 458 | ||
| NM_001271484.1 | NP_001258413.1 | 4 | 543 | |||
| NM_030179.3 | NP_084455.2 | 4 | 446 | |||
| NM_175378.1 | NP_780587.1 | 4 | 543 | |||
| GenBank mRNA | AK017914.1 | - | 1 | 33 | ||
| AK018895.1 | - | 4 | 536 | |||
| AK019516.1 | - | 4 | 459 | |||
| AK029962.1 | - | 4 | 544 | |||
| AK075758.1 | - | 4 | 534 | |||
| AK161301.1 | - | 4 | 533 | |||
| BC035226.1 | - | 4 | 446 | 4920788 | ||

Clip4
Genomic Map