siRNA Details
| Gene Symbol: Usp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030180.2 | NP_084456.2 | 21 | 5033 | ||
| GenBank mRNA | AK019588.1 | - | 4 | 1220 | ||
| BC066166.1 | - | 4 | 1544 | |||
| BC066175.1 | - | 4 | 1544 | |||
| BC117844.1 | - | 4 | 1208 | |||
| BC117845.1 | - | 4 | 1208 | |||

Usp54
Genomic Map